ENA375227620 Enamine

1-(1-bromoethyl)-2-chloro-4,5-dimethoxybenzene

  • MDL number MFCD12790266
  • Cas Number : 1248231-47-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-114867

Safety Information

msds https://www.enaminestore.com/catalog/EN300-114867
Availability details for
Quantity
  • Availability
  • Add to Cart