ENA375227624 Enamine

1-(2,2-diethoxyethyl)-4-methoxypiperidine

  • MDL number MFCD18848405
  • Cas Number : 1343614-00-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-114872

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-114872
Availability details for
Quantity
  • Availability
  • Add to Cart