ENA375227627 Enamine

2-{7,9-dimethyl-2,4-dioxo-1,3-diazaspiro[4.5]decan-3-yl}acetic acid

  • MDL number MFCD20600336
  • Cas Number : 1378433-28-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 259...261
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-113548

Safety Information

msds https://www.enaminestore.com/catalog/EN300-113548
Availability details for
Quantity
  • Availability
  • Add to Cart