ENA375227629 Enamine

(6-fluoropyridin-2-yl)methanamine hydrochloride

  • MDL number MFCD19982295
  • Cas Number : 1423025-98-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 152...154
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-114878

Safety Information

msds https://www.enaminestore.com/catalog/EN300-114878
Availability details for
Quantity
  • Availability
  • Add to Cart