ENA375227631 Enamine

2-formyl-3-methylbutanenitrile

  • MDL number MFCD14621602
  • Cas Number : 771510-15-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-114882

Safety Information

msds https://www.enaminestore.com/catalog/EN300-114882
Availability details for
Quantity
  • Availability
  • Add to Cart