ENA375227633 Enamine

2-methyl-2-(methylsulfanyl)propanenitrile

  • MDL number MFCD21127166
  • Cas Number : 10074-86-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-114885

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-114885
Availability details for
Quantity
  • Availability
  • Add to Cart