ENA375227634 Enamine

2-(4-methylpiperazin-1-yl)-1-[4-(propan-2-yloxy)phenyl]ethan-1-ol

  • MDL number MFCD19277678
  • Cas Number : 1267705-78-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-114886

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-114886
Availability details for
Quantity
  • Availability
  • Add to Cart