ENA375227641 Enamine

(2-methyl-1,3-dioxolan-2-yl)methanamine

  • MDL number MFCD10698199
  • Cas Number : 3289-19-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-114894

Safety Information

signalword Danger
msds https://www.enaminestore.com/catalog/EN300-114894
pictograms GHS05,GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart