ENA375227652 Enamine

3-(butan-2-yl)-1-[(6-methylpyridin-3-yl)methyl]piperazine

  • MDL number MFCD21452113
  • Cas Number : 1423024-11-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-114908

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-114908
Availability details for
Quantity
  • Availability
  • Add to Cart