ENA375227653 Enamine

1-cyclopropylpent-4-en-1-one

  • MDL number MFCD18336725
  • Cas Number : 31594-49-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-114912

Safety Information

msds https://www.enaminestore.com/catalog/EN300-114912
Availability details for
Quantity
  • Availability
  • Add to Cart