ENA375227654 Enamine

3-[(3-fluorophenyl)methyl]pyrrolidin-3-ol

  • MDL number MFCD18368662
  • Cas Number : 1246732-83-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 59...61
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-114913

Safety Information

msds https://www.enaminestore.com/catalog/EN300-114913
Availability details for
Quantity
  • Availability
  • Add to Cart