ENA375227656 Enamine

2-amino-4,4-dimethylpentanoic acid hydrochloride

  • MDL number MFCD09033881
  • Cas Number : 57224-48-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-114920

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-114920
Availability details for
Quantity
  • Availability
  • Add to Cart