ENA375227662 Enamine

4-(hydroxymethyl)-3-methylbenzonitrile

  • MDL number MFCD18394344
  • Cas Number : 227094-07-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 66...68
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-114948

Safety Information

signalword Danger
msds https://www.enaminestore.com/catalog/EN300-114948
pictograms GHS05,GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart