ENA375227663 Enamine

4-(dimethylamino)-3-fluorobenzonitrile

  • MDL number MFCD12110121
  • Cas Number : 160658-69-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-114949

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-114949
Availability details for
Quantity
  • Availability
  • Add to Cart