ENA375227666 Enamine

methyl imidazo[1,5-a]pyridine-7-carboxylate

  • MDL number MFCD20526488
  • Cas Number : 1377829-50-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-113912

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-113912
Availability details for
Quantity
  • Availability
  • Add to Cart