ENA375227668 Enamine

2,3-dimethyl piperidine-2,3-dicarboxylate

  • MDL number MFCD12026413
  • Cas Number : 23580-75-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-114957

Safety Information

msds https://www.enaminestore.com/catalog/EN300-114957
Availability details for
Quantity
  • Availability
  • Add to Cart