ENA375227670 Enamine

N-[(1-aminocyclohexyl)methyl]-1H-pyrrole-3-sulfonamide

  • MDL number MFCD12083377
  • Cas Number : 1153056-62-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-114959

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-114959
Availability details for
Quantity
  • Availability
  • Add to Cart