ENA375227687 Enamine

1-(2-bromoethyl)-3,5-dichlorobenzene

  • MDL number MFCD20432741
  • Cas Number : 93427-14-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-114996

Safety Information

msds https://www.enaminestore.com/catalog/EN300-114996
Availability details for
Quantity
  • Availability
  • Add to Cart