ENA375227691 Enamine

1-(2-bromoethoxy)-3-methoxybenzene

  • MDL number MFCD00980056
  • Cas Number : 3245-45-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-113976

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-113976
Availability details for
Quantity
  • Availability
  • Add to Cart