ENA375227695 Enamine

[4-(aminomethyl)oxan-4-yl]methanol

  • MDL number MFCD09864391
  • Cas Number : 959238-22-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 54...56
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-115011

Safety Information

msds https://www.enaminestore.com/catalog/EN300-115011
Availability details for
Quantity
  • Availability
  • Add to Cart