ENA375227707 Enamine

(2-amino-2-methylpropoxy)trimethylsilane

  • MDL number MFCD09840503
  • Cas Number : 38421-19-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-115057

Safety Information

msds https://www.enaminestore.com/catalog/EN300-115057
Availability details for
Quantity
  • Availability
  • Add to Cart