ENA375227709 Enamine

4,4-dimethylcyclohexane-1-sulfonyl chloride

  • MDL number MFCD20306414
  • Cas Number : 1344070-27-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -10
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-115059

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-115059
Availability details for
Quantity
  • Availability
  • Add to Cart