ENA375227721 Enamine

1,2,3,4-tetrahydroquinoline-7-sulfonic acid

  • MDL number MFCD09839673
  • Cas Number : 94411-91-3
  • Molecular Weight : 213.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-115100

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-115100
Availability details for
Quantity
  • Availability
  • Add to Cart