ENA375227722 Enamine

(4-aminobutyl)(methyl)amine

  • MDL number MFCD12783777
  • Cas Number : 14475-60-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-115101

Safety Information

msds https://www.enaminestore.com/catalog/EN300-115101
Availability details for
Quantity
  • Availability
  • Add to Cart