ENA375227723 Enamine

2,3-dihydro-1-benzofuran-3-carbonyl chloride

  • MDL number MFCD19657281
  • Cas Number : 1343949-08-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-115102

Safety Information

msds https://www.enaminestore.com/catalog/EN300-115102
Availability details for
Quantity
  • Availability
  • Add to Cart