ENA375227732 Enamine

3-amino-N-[(thiophen-2-yl)methyl]benzamide

  • MDL number MFCD11650816
  • Cas Number : 1094671-91-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-115133

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-115133
Availability details for
Quantity
  • Availability
  • Add to Cart