ENA375227734 Enamine

propyl 4-chlorobenzoate

  • MDL number MFCD12178053
  • Cas Number : 25800-30-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-115135

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-115135
Availability details for
Quantity
  • Availability
  • Add to Cart