ENA375227736 Enamine

methyl 3-(2-phenoxypropanamido)propanoate

  • MDL number MFCD13223814
  • Cas Number : 1306138-32-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-115137

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-115137
Availability details for
Quantity
  • Availability
  • Add to Cart