ENA375227740 Enamine

2-amino-N-methyl-N-[(5-methyl-1,2-oxazol-3-yl)methyl]-2-phenylacetamide

  • MDL number MFCD12484723
  • Cas Number : 1218740-86-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-115142

Safety Information

msds https://www.enaminestore.com/catalog/EN300-115142
Availability details for
Quantity
  • Availability
  • Add to Cart