ENA375227756 Enamine

2-cyano-3-methylbenzene-1-sulfonyl chloride

  • MDL number MFCD18394316
  • Cas Number : 850154-02-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 75-77
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-115165

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-115165
Availability details for
Quantity
  • Availability
  • Add to Cart