ENA375227758 Enamine

3-methyl-2-[4-(trifluoromethyl)phenyl]pyrrolidine

  • MDL number MFCD05189877
  • Cas Number : 526182-73-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-114201

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-114201
Availability details for
Quantity
  • Availability
  • Add to Cart