ENA375227761 Enamine

N-(2-methoxyphenyl)-2-(piperazin-1-yl)acetamide hydrochloride

  • MDL number MFCD05656290
  • Cas Number : 1158358-15-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-115188

Safety Information

msds https://www.enaminestore.com/catalog/EN300-115188
Availability details for
Quantity
  • Availability
  • Add to Cart