ENA375227764 Enamine

benzyl N-(2-oxocyclobutyl)carbamate

  • MDL number MFCD22208485
  • Cas Number : 406951-43-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-108330

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-108330
Availability details for
Quantity
  • Availability
  • Add to Cart