ENA375227768 Enamine

(naphthalen-1-yl)urea

  • MDL number MFCD00021444
  • Cas Number : 6950-84-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 284...286
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-115215

Safety Information

signalword Danger
msds https://www.enaminestore.com/catalog/EN300-115215
pictograms GHS06
Availability details for
Quantity
  • Availability
  • Add to Cart