ENA375227778 Enamine

2-(3-methoxyphenyl)-2-methylpropan-1-amine

  • MDL number MFCD09414706
  • Cas Number : 60812-46-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-115229

Safety Information

msds https://www.enaminestore.com/catalog/EN300-115229
Availability details for
Quantity
  • Availability
  • Add to Cart