ENA375227779 Enamine

3-(3-chloro-4,5-dimethoxyphenyl)-5-(chloromethyl)-1,2,4-oxadiazole

  • MDL number MFCD09051408
  • Cas Number : 937630-31-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-108415

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-108415
Availability details for
Quantity
  • Availability
  • Add to Cart