ENA375227786 Enamine

2-bromo-3,6-dichloropyridine

  • MDL number MFCD18447566
  • Cas Number : 1266119-41-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-115245

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-115245
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart