ENA375227790 Enamine

6-hydroxy-2-methylpyridine-3-carboxylic acid

  • MDL number MFCD09881218
  • Cas Number : 66909-37-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 329...331
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-114264

Safety Information

msds https://www.enaminestore.com/catalog/EN300-114264
Availability details for
Quantity
  • Availability
  • Add to Cart