ENA375227791 Enamine

3-(oxolan-3-yl)pyrrolidine

  • MDL number MFCD16707136
  • Cas Number : 1121057-47-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-115251

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-115251
Availability details for
Quantity
  • Availability
  • Add to Cart