ENA375227794 Enamine

3-(dimethylamino)benzene-1-sulfonyl chloride

  • MDL number MFCD19200075
  • Cas Number : 876482-47-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-115254

Safety Information

msds https://www.enaminestore.com/catalog/EN300-115254
Availability details for
Quantity
  • Availability
  • Add to Cart