ENA375227800 Enamine

2-[4-(methylsulfanyl)phenyl]-2-oxoacetaldehyde

  • MDL number MFCD22565974
  • Cas Number : 58950-88-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-115293

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-115293
Availability details for
Quantity
  • Availability
  • Add to Cart