ENA375227806 Enamine

methyl 3-methyl-3-(methylamino)butanoate

  • MDL number MFCD19222840
  • Cas Number : 89855-40-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-115299

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-115299
Availability details for
Quantity
  • Availability
  • Add to Cart