ENA375227808 Enamine

1-(2-aminoethoxy)-2-chlorobenzene

  • MDL number MFCD00125294
  • Cas Number : 26378-53-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-108477

Safety Information

signalword Danger
msds https://www.enaminestore.com/catalog/EN300-108477
pictograms GHS05
Availability details for
Quantity
  • Availability
  • Add to Cart