ENA375227811 Enamine

2-chloro-5-[(dimethylcarbamoyl)amino]benzoic acid

  • MDL number MFCD14596153
  • Cas Number : 1250075-49-0
  • Molecular Weight : 242.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-115305

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-115305
Availability details for
Quantity
  • Availability
  • Add to Cart