ENA375227815 Enamine

3-(cyclopropylmethyl)-1-phenyl-1H-pyrazol-5-amine

  • MDL number MFCD16665739
  • Cas Number : 1250517-20-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-115311

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-115311
Availability details for
Quantity
  • Availability
  • Add to Cart