ENA375227823 Enamine

3-[(1H-indazol-5-yl)carbamoyl]prop-2-enoic acid

  • MDL number MFCD02656565
  • Cas Number : 332033-19-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-115326

Safety Information

msds https://www.enaminestore.com/catalog/EN300-115326
Availability details for
Quantity
  • Availability
  • Add to Cart