ENA375227832 Enamine

(4-methoxypyridin-2-yl)methanamine dihydrochloride

  • MDL number MFCD20327973
  • Cas Number : 1344046-06-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 169...171
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-115348

Safety Information

msds https://www.enaminestore.com/catalog/EN300-115348
Availability details for
Quantity
  • Availability
  • Add to Cart