ENA375227833 Enamine

3-[5-methyl-2-(propan-2-yl)phenoxy]propanoic acid

  • MDL number MFCD09045505
  • Cas Number : 97395-13-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-114334

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-114334
Availability details for
Quantity
  • Availability
  • Add to Cart