ENA375227834 Enamine

1-cyclopropylpent-4-en-1-ol

  • MDL number MFCD18337821
  • Cas Number : 1411260-56-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-115353

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-115353
Availability details for
Quantity
  • Availability
  • Add to Cart