ENA375227838 Enamine

2-amino-3-methyl-N-(2-methylphenyl)pentanamide

  • MDL number MFCD09947727
  • Cas Number : 1104197-17-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-115357

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-115357
Availability details for
Quantity
  • Availability
  • Add to Cart